Anti-Angiotensin II/III Mouse Monoclonal Antibody (DyLight 550) [clone: Ang II E7 (BGN/KA/4L)]

Supplier: Novus Biologicals
NB100-62346R
103413-142EA 706.28 USD
103413-142
Anti-Angiotensin II/III Mouse Monoclonal Antibody (DyLight 550) [clone: Ang II E7 (BGN/KA/4L)]
Antibodies
The Angiotensin II / III Antibody (Ang II E7 (BGN / KA / 4L)) [DyLight 550] from Novus Biologicals is a mouse monoclonal antibody to Angiotensin II / III. This antibody reacts with human, mouse, rat. The Angiotensin II / III Antibody (Ang II E7 (BGN / KA / 4L)) [DyLight 550] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Angiotensin II/III
Clonality: Monoclonal
Clone: Ang II E7 (BGN/KA/4L)
Conjugation: DyLight 550
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR