Anti-Angiotensin I Mouse Monoclonal Antibody (DyLight 405) [clone: BGN/KA/4H]

Supplier: Novus Biologicals
NB100-62344V
103413-202EA 706.28 USD
103413-202
Anti-Angiotensin I Mouse Monoclonal Antibody (DyLight 405) [clone: BGN/KA/4H]
Antibodies
The Angiotensin I Antibody (BGN / KA / 4H) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to Angiotensin I. This antibody reacts with human. The Angiotensin I Antibody (BGN / KA / 4H) [DyLight 405] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Angiotensin I
Clonality: Monoclonal
Clone: BGN/KA/4H
Conjugation: DyLight 405
Epitope: P-F-H-L-V-I-H-N-E-S(-C)
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR