Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Amodiaquine Mouse Monoclonal Antibody (DyLight 755) [clone: 6D10]

Supplier: Novus Biologicals
NBP1-05106IR
The Amodiaquine Antibody (6D10) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to Amodiaquine. This antibody reacts with non-species specific. The Amodiaquine Antibody (6D10) [DyLight 755] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Amodiaquine
Clonality: Monoclonal
Clone: 6D10
Conjugation: DyLight 755
Epitope: Specific for the non-quinoline sidechain of amodiaquine
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Non-species specific




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-05106IR 103410-862 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Name Amodiaquine
Reactivity Non-species specific
Conjugation DyLight® 755
Clonality Monoclonal
Antibody Type Primary
Clone 6D10
Host Mouse
Isotype IgG2a Kappa
ELISA Yes
Epitope Specific for the non-quinoline sidechain of amodiaquine
Immunogen 4-amino-2(diethylaminomethyl)phenol a preparation of a carrier protein
Purification Protein A purified
Storage Buffer 50mM Sodium Borate
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty