Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Amodiaquine Mouse Monoclonal Antibody (DyLight 488) [clone: 6D10]

Supplier: Novus Biologicals
NBP1-05106G
The Amodiaquine Antibody (6D10) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to Amodiaquine. This antibody reacts with non-species specific. The Amodiaquine Antibody (6D10) [DyLight 488] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Amodiaquine
Clonality: Monoclonal
Clone: 6D10
Conjugation: DyLight 488
Epitope: Specific for the non-quinoline sidechain of amodiaquine
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Non-species specific




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-05106G 103410-854 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Name Amodiaquine
Reactivity Non-species specific
Conjugation DyLight® 488
Clonality Monoclonal
Antibody Type Primary
Clone 6D10
Host Mouse
Isotype IgG2a Kappa
ELISA Yes
Epitope Specific for the non-quinoline sidechain of amodiaquine
Immunogen 4-amino-2(diethylaminomethyl)phenol a preparation of a carrier protein
Purification Protein A purified
Storage Buffer 50mM Sodium Borate
Storage Temperature Store at 4C in the dark.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty