Anti-Amodiaquine Mouse Monoclonal Antibody (Biotin) [clone: 6D10]

Supplier: Novus Biologicals
NBP1-05106B
103410-846EA 399 USD
103410-846
Anti-Amodiaquine Mouse Monoclonal Antibody (Biotin) [clone: 6D10]
The Amodiaquine Antibody (6D10) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to Amodiaquine. This antibody reacts with non-species specific. The Amodiaquine Antibody (6D10) [Biotin] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Amodiaquine
Clonality: Monoclonal
Clone: 6D10
Conjugation: Biotin
Epitope: Specific for the non-quinoline sidechain of amodiaquine
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR