Anti-alpha-Synuclein Chicken Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-25146
103405-804EA 491.08 USD
103405-804
Anti-alpha-Synuclein Chicken Polyclonal Antibody
Antibodies
The alpha-Synuclein Antibody from Novus Biologicals is a chicken polyclonal antibody to alpha-Synuclein. This antibody reacts with human, rat. The alpha-Synuclein Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: alpha-Synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR