Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1A4 + ACTA2/791]

Supplier: Novus Biologicals
NBP2-44464F
103410-682
Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1A4 + ACTA2/791]
The alpha-Smooth Muscle Actin Antibody (1A4 + ACTA2 / 791) [FITC] from Novus Biologicals is a mouse monoclonal antibody to alpha-Smooth Muscle Actin. This antibody reacts with human, rat. The alpha-Smooth Muscle Actin Antibody (1A4 + ACTA2 / 791) [FITC] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: alpha-Smooth Muscle Actin
Clonality: Monoclonal
Clone: 1A4 + ACTA2/791
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty