Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (DyLight 680) [clone: SPM332]

Supplier: Novus Biologicals
NBP2-34760FR
103410-706EA 638.77 USD
103410-706
Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (DyLight 680) [clone: SPM332]
Antibodies
The alpha-Smooth Muscle Actin Antibody (SPM332) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to alpha-Smooth Muscle Actin. This antibody reacts with human, mouse, rat, porcine, baboon, bovine, canine, chicken, feline, guinea pig, goat, primate, rabbit, sheep. The alpha-Smooth Muscle Actin Antibody (SPM332) [DyLight 680] has been validated for the following applications: Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: alpha-Smooth Muscle Actin
Clonality: Monoclonal
Clone: SPM332
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat, Porcine, Baboon, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Primate, Rabbit, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR