Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: ACTA2/791]

Supplier: Novus Biologicals
NBP2-47698H
103410-748EA 638.77 USD
103410-748
Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: ACTA2/791]
Antibodies
The alpha-Smooth Muscle Actin Antibody (ACTA2 / 791) [HRP] from Novus Biologicals is a mouse monoclonal antibody to alpha-Smooth Muscle Actin. This antibody reacts with human, rat. The alpha-Smooth Muscle Actin Antibody (ACTA2 / 791) [HRP] has been validated for the following applications: Immunohistochemistry-Paraffin.

Type: Primary
Antigen: alpha-Smooth Muscle Actin
Clonality: Monoclonal
Clone: ACTA2/791
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR