Anti-alpha-Sarcoglycan Mouse Monoclonal Antibody [clone: 3C4]

Supplier: Novus Biologicals
H00006442-M01
103310-066EA 798.46 USD
103310-066
Anti-alpha-Sarcoglycan Mouse Monoclonal Antibody [clone: 3C4]
Antibodies
The alpha-Sarcoglycan Antibody (3C4) from Novus Biologicals is a mouse monoclonal antibody to alpha-Sarcoglycan. This antibody reacts with human. The alpha-Sarcoglycan Antibody (3C4) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: alpha-Sarcoglycan
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR