Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (Biotin) [clone: AAP1]

Supplier: Novus Biologicals
NB100-2638B
103412-592EA 614.15 USD
103412-592
Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (Biotin) [clone: AAP1]
Antibodies
The Alkaline Phosphatase, Intestinal Antibody (AAP1) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to Alkaline Phosphatase, Intestinal. This antibody reacts with human. The Alkaline Phosphatase, Intestinal Antibody (AAP1) [Biotin] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Alkaline Phosphatase, Intestinal
Clonality: Monoclonal
Clone: AAP1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR