Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AP1B9]

Supplier: Novus Biologicals
NB100-2637F
103412-604EA 552.62 USD
103412-604
Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AP1B9]
Antibodies
The Alkaline Phosphatase, Intestinal Antibody (AP1B9) [FITC] from Novus Biologicals is a mouse monoclonal antibody to Alkaline Phosphatase, Intestinal. This antibody reacts with human, bovine. The Alkaline Phosphatase, Intestinal Antibody (AP1B9) [FITC] has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Alkaline Phosphatase, Intestinal
Clonality: Monoclonal
Clone: AP1B9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR