Anti-Alkaline Phosphatase, Intestinal Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-33624
103283-132EA 734.58 USD
103283-132
Anti-Alkaline Phosphatase, Intestinal Rabbit Polyclonal Antibody
Antibodies
The Alkaline Phosphatase, Intestinal Antibody from Novus Biologicals is a rabbit polyclonal antibody to Alkaline Phosphatase, Intestinal. This antibody reacts with human. The Alkaline Phosphatase, Intestinal Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Alkaline Phosphatase, Intestinal
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR