Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3F11]

Supplier: Novus Biologicals
NBP1-05093F
103412-582EA 399 USD
103412-582
Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3F11]
The Alkaline Phosphatase, Intestinal Antibody (3F11) [FITC] from Novus Biologicals is a mouse monoclonal antibody to Alkaline Phosphatase, Intestinal. This antibody reacts with bovine. The Alkaline Phosphatase, Intestinal Antibody (3F11) [FITC] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Alkaline Phosphatase, Intestinal
Clonality: Monoclonal
Clone: 3F11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR