Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (Biotin) [clone: 3F11]

Supplier: Novus Biologicals
NBP1-05093B
103410-140EA 399 USD
103410-140
Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (Biotin) [clone: 3F11]
The Alkaline Phosphatase, Intestinal Antibody (3F11) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to Alkaline Phosphatase, Intestinal. This antibody reacts with bovine. The Alkaline Phosphatase, Intestinal Antibody (3F11) [Biotin] has been validated for the following applications: ELISA.

Type: Primary
Antigen: Alkaline Phosphatase, Intestinal
Clonality: Monoclonal
Clone: 3F11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR