Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (DyLight 405) [clone: SPM372]

Supplier: Novus Biologicals
NBP2-47995V
103412-618EA 638.77 USD
103412-618
Anti-Alkaline Phosphatase, Intestinal Mouse Monoclonal Antibody (DyLight 405) [clone: SPM372]
Antibodies
The Alkaline Phosphatase, Intestinal Antibody (SPM372) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to Alkaline Phosphatase, Intestinal. This antibody reacts with human, primate. The Alkaline Phosphatase, Intestinal Antibody (SPM372) [DyLight 405] has been validated for the following applications: Flow Cytometry, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Alkaline Phosphatase, Intestinal
Clonality: Monoclonal
Clone: SPM372
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR