Anti-CCL20/MIP-3 alpha Mouse Monoclonal Antibody (Biotin) [clone: 308B7.06]

Supplier: Novus Biologicals
DDX0431B-100
103318-968EA 777.05 USD
103318-968
Anti-CCL20/MIP-3 alpha Mouse Monoclonal Antibody (Biotin) [clone: 308B7.06]
Antibodies
The CCL20 / MIP-3 alpha Antibody (308B7.06) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to CCL20 / MIP-3 alpha. This antibody reacts with human. The CCL20 / MIP-3 alpha Antibody (308B7.06) [Biotin] has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry.

Type: Primary
Antigen: CCL20/MIP-3 alpha
Clonality: Monoclonal
Clone: 308B7.06
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR