Anti-CCL18/PARC Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00006362-D01P
103309-920EA 429 USD
103309-920
Anti-CCL18/PARC Rabbit Polyclonal Antibody
Antibodies
The CCL18 / PARC Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCL18 / PARC. This antibody reacts with human. The CCL18 / PARC Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CCL18/PARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR