Anti-CCL15/MIP-1 delta Mouse Monoclonal Antibody [clone: 3B7]

Supplier: Novus Biologicals
H00006359-M04
103309-914EA 767.69 USD
103309-914
Anti-CCL15/MIP-1 delta Mouse Monoclonal Antibody [clone: 3B7]
Antibodies
The CCL15 / MIP-1 delta Antibody (3B7) from Novus Biologicals is a mouse monoclonal antibody to CCL15 / MIP-1 delta. This antibody reacts with human. The CCL15 / MIP-1 delta Antibody (3B7) has been validated for the following applications: ELISA.

Type: Primary
Antigen: CCL15/MIP-1 delta
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR