Anti-CCL13/MCP-4 Mouse Monoclonal Antibody [clone: S2]

Supplier: Novus Biologicals
H00006357-M03
103309-898EA 399 USD
103309-898
Anti-CCL13/MCP-4 Mouse Monoclonal Antibody [clone: S2]
Antibodies
The CCL13 / MCP-4 Antibody (S2) from Novus Biologicals is a mouse monoclonal antibody to CCL13 / MCP-4. This antibody reacts with human. The CCL13 / MCP-4 Antibody (S2) has been validated for the following applications: ELISA.

Type: Primary
Antigen: CCL13/MCP-4
Clonality: Monoclonal
Clone: S2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR