Anti-CCL11/Eotaxin Mouse Monoclonal Antibody [clone: 1C8-2C1]

Supplier: Novus Biologicals
H00006356-M01
103309-890EA 798.46 USD
103309-890
Anti-CCL11/Eotaxin Mouse Monoclonal Antibody [clone: 1C8-2C1]
Antibodies
The CCL11 / Eotaxin Antibody (1C8-2C1) from Novus Biologicals is a mouse monoclonal antibody to CCL11 / Eotaxin. This antibody reacts with human. The CCL11 / Eotaxin Antibody (1C8-2C1) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CCL11/Eotaxin
Clonality: Monoclonal
Clone: 1C8-2C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR