Anti-CCDC94 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91766
103278-352EA 734.58 USD
103278-352
Anti-CCDC94 Rabbit Polyclonal Antibody
Antibodies
The CCDC94 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCDC94. This antibody reacts with human, mouse. The CCDC94 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CCDC94
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR