Anti-CCDC90A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15755
102706-604EA 635.51 USD
102706-604
Anti-CCDC90A Rabbit Polyclonal Antibody
Antibodies
The CCDC90A Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCDC90A. This antibody reacts with mouse. The CCDC90A Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CCDC90A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR