Anti-Cav3.2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88176
103271-036EA 734.58 USD
103271-036
Anti-Cav3.2 Rabbit Polyclonal Antibody
Antibodies
The Cav3.2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cav3.2. This antibody reacts with human. The Cav3.2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Cav3.2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR