Anti-Cav1.3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86684
103270-322EA 734.58 USD
103270-322
Anti-Cav1.3 Rabbit Polyclonal Antibody
Antibodies
The Cav1.3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cav1.3. This antibody reacts with human. The Cav1.3 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Cav1.3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR