Anti-Cav1.2 Mouse Monoclonal Antibody [clone: 4D10]

Supplier: Novus Biologicals
H00000775-M20
103322-994EA 798.46 USD
103322-994
Anti-Cav1.2 Mouse Monoclonal Antibody [clone: 4D10]
Antibodies
The Cav1.2 Antibody (4D10) from Novus Biologicals is a mouse monoclonal antibody to Cav1.2. This antibody reacts with human. The Cav1.2 Antibody (4D10) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Cav1.2
Clonality: Monoclonal
Clone: 4D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR