Anti-CATSPERB Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15728
102706-552EA 635.51 USD
102706-552
Anti-CATSPERB Rabbit Polyclonal Antibody
Antibodies
The CATSPERB Antibody from Novus Biologicals is a rabbit polyclonal antibody to CATSPERB. This antibody reacts with human. The CATSPERB Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CATSPERB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR