Anti-CAT1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90066
103277-524EA 734.58 USD
103277-524
Anti-CAT1 Rabbit Polyclonal Antibody
Antibodies
The CAT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CAT1. This antibody reacts with human. The CAT1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR