Anti-CASZ1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86617
103273-800EA 734.58 USD
103273-800
Anti-CASZ1 Rabbit Polyclonal Antibody
Antibodies
The CASZ1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CASZ1. This antibody reacts with human. The CASZ1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunocytochemistry.

Type: Primary
Antigen: CASZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR