Anti-Caspr2 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00026047-B01P
103342-040EA 399 USD
103342-040
Anti-Caspr2 Mouse Polyclonal Antibody
Antibodies
The Caspr2 Antibody from Novus Biologicals is a mouse polyclonal antibody to Caspr2. This antibody reacts with human. The Caspr2 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Caspr2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR