Anti-CART1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88190
103271-062EA 734.58 USD
103271-062
Anti-CART1 Rabbit Polyclonal Antibody
Antibodies
The CART1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CART1. This antibody reacts with human. The CART1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CART1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR