Anti-CARKL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91151
103279-114EA 734.58 USD
103279-114
Anti-CARKL Rabbit Polyclonal Antibody
Antibodies
The CARKL Antibody from Novus Biologicals is a rabbit polyclonal antibody to CARKL. This antibody reacts with human. The CARKL Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CARKL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR