Anti-CARHSP1 Mouse Monoclonal Antibody [clone: 4G10]

Supplier: Novus Biologicals
H00023589-M08
103341-568EA 798.46 USD
103341-568
Anti-CARHSP1 Mouse Monoclonal Antibody [clone: 4G10]
Antibodies
The CARHSP1 Antibody (4G10) from Novus Biologicals is a mouse monoclonal antibody to CARHSP1. This antibody reacts with human. The CARHSP1 Antibody (4G10) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Recommended Dilutions: Western Blot: 1 to 5 µg/ml.

Type: Primary
Antigen: CARHSP1
Clonality: Monoclonal
Clone: 4G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR