Anti-CARF/CDKN2AIP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86628
103273-814EA 734.58 USD
103273-814
Anti-CARF/CDKN2AIP Rabbit Polyclonal Antibody
Antibodies
The CARF / CDKN2AIP Antibody from Novus Biologicals is a rabbit polyclonal antibody to CARF / CDKN2AIP. This antibody reacts with human. The CARF / CDKN2AIP Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CARF/CDKN2AIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR