Anti-CARD6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56111
103358-536EA 229 USD
103358-536
Anti-CARD6 Rabbit Polyclonal Antibody
Antibodies
The CARD6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CARD6. This antibody reacts with human. The CARD6 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CARD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR