Anti-CARD12 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56143
103358-630EA 229 USD
103358-630
Anti-CARD12 Rabbit Polyclonal Antibody
Antibodies
The CARD12 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CARD12. This antibody reacts with human. The CARD12 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CARD12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR