Anti-CAPN9 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-14896
102704-994EA 635.51 USD
102704-994
Anti-CAPN9 Rabbit Polyclonal Antibody
Antibodies
The Calpain 9 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calpain 9. This antibody reacts with human. The Calpain 9 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CAPN9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR