Anti-CAPN2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-88204
103271-086EA 734.58 USD
103271-086
Anti-CAPN2 Rabbit Polyclonal Antibody
Antibodies
The Calpain 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calpain 2. This antibody reacts with human, mouse, rat. The Calpain 2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CAPN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR