Anti-CAPN2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00000824-D01P
103323-128EA 429 USD
103323-128
Anti-CAPN2 Rabbit Polyclonal Antibody
Antibodies
The Calpain 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calpain 2. This antibody reacts with human. The Calpain 2 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CAPN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR