Anti-CAPN15 Mouse Monoclonal Antibody [clone: 4E2.]

Supplier: Novus Biologicals
H00006650-M05
103310-508EA 399 USD
103310-508
Anti-CAPN15 Mouse Monoclonal Antibody [clone: 4E2.]
Antibodies
The Calpain 15 Antibody (4E2.) from Novus Biologicals is a mouse monoclonal antibody to Calpain 15. This antibody reacts with human. The Calpain 15 Antibody (4E2.) has been validated for the following applications: ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CAPN15
Clonality: Monoclonal
Clone: 4E2.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR