Anti-CAPN11 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-33942
103288-420EA 734.58 USD
103288-420
Anti-CAPN11 Rabbit Polyclonal Antibody
Antibodies
The Calpain 11 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calpain 11. This antibody reacts with human. The Calpain 11 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CAPN11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR