Anti-CAPN10 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00011132-D01P
103340-206EA 844.62 USD
103340-206
Anti-CAPN10 Rabbit Polyclonal Antibody
Antibodies
The Calpain 10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calpain 10. This antibody reacts with human. The Calpain 10 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CAPN10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR