Anti-CAPN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15671
102706-442EA 635.51 USD
102706-442
Anti-CAPN1 Rabbit Polyclonal Antibody
Antibodies
The Calpain 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calpain 1. This antibody reacts with human, mouse. The Calpain 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: CAPN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR