Anti-CAPN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00000823-D01P
103323-126EA 429 USD
103323-126
Anti-CAPN1 Rabbit Polyclonal Antibody
Antibodies
The Calpain 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Calpain 1. This antibody reacts with human. The Calpain 1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Proximity Ligation Assay.

Type: Primary
Antigen: CAPN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR