Anti-capg-1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
48460002-0.1MG
103317-890EA 549 USD
103317-890
Anti-capg-1 Rabbit Polyclonal Antibody
Antibodies
The capg-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to capg-1. This antibody reacts with c. elegans. The capg-1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: capg-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: MPPKKRIRGPKLPALREEKSTGTDVASDESFNESADFLNNEELNNDQNDAENTVLSEGVSTLRINENMTKEDRACISAALFIKKRVVESIRTVFQSRDDI
Host: Rabbit
Isotype: IgG
Reactivity: C. elegans
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR