Anti-CAP2 Mouse Monoclonal Antibody [clone: 3G9-1A5]

Supplier: Novus Biologicals
H00010486-M01
103338-996EA 798.46 USD
103338-996
Anti-CAP2 Mouse Monoclonal Antibody [clone: 3G9-1A5]
Antibodies
The CAP2 Antibody (3G9-1A5) from Novus Biologicals is a mouse monoclonal antibody to CAP2. This antibody reacts with human. The CAP2 Antibody (3G9-1A5) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CAP2
Clonality: Monoclonal
Clone: 3G9-1A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR