Anti-CAP1 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00010487-B01P
103338-998EA 399 USD
103338-998
Anti-CAP1 Mouse Polyclonal Antibody
Antibodies
The CAP1 Antibody from Novus Biologicals is a mouse polyclonal antibody to CAP1. This antibody reacts with human. The CAP1 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR