Anti-CaMKK2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-15688
102706-472EA 349 USD
102706-472
Anti-CaMKK2 Rabbit Polyclonal Antibody
The CaMKK2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CaMKK2. This antibody reacts with human, mouse. The CaMKK2 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CaMKK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR