Anti-CaMKK alpha Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00084254-D01P
103345-128EA 429 USD
103345-128
Anti-CaMKK alpha Rabbit Polyclonal Antibody
Antibodies
The CaMKK alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to CaMKK alpha. This antibody reacts with human. The CaMKK alpha Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: CaMKK alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR