Anti-CaMKIV Mouse Monoclonal Antibody [clone: 1A3]

Supplier: Novus Biologicals
H00000814-M01
103323-072EA 798.46 USD
103323-072
Anti-CaMKIV Mouse Monoclonal Antibody [clone: 1A3]
Antibodies
The CaMKIV Antibody (1A3) from Novus Biologicals is a mouse monoclonal antibody to CaMKIV. This antibody reacts with human. The CaMKIV Antibody (1A3) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, RNA Inhibition.

RNA Inhibition

Type: Primary
Antigen: CaMKIV
Clonality: Monoclonal
Clone: 1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR